Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water Solution, 0.1%(v/v) Trifluoroacetic Acid, for HPLC, Solstice

Catalog No. 60047120 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-120 4 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-047-120 Supplier Solstice Supplier No. 5769054X4L

missing translation for 'validMainframeMsg'

CHROMASOLV™, LabReady™ Solvent Blend, contains 0.1% (v/v) trifluoroacetic acid, for HPLC

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-05-1 , 7732-18-5
MDL Number MFCD00004169
UN Number NONH for all modes of transport
Chemical Name or Material Water solution

Specifications

Color Colorless
Density 4.930g/mL
Quantity 4 x 4 L
Packaging Glass Bottle
Physical Form Liquid
Molecular Formula C2HF3O2
Linear Formula H2O
Boiling Point 100°C
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
SMILES OC(=O)C(F)(F)F
IUPAC Name trifluoroacetic acid
Molecular Weight (g/mol) 114.02
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.