Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water Solution contains 20 (v/v) acetonitrile, 0.1 (v/v) formic acid, MilliporeSigma™ Supelco™

Catalog No. 111016693 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
4 x 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-6693 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-6693 Supplier MilliporeSigma Supelco Supplier No. 6333214X4L
Add to Cart
Add to Cart
The product is water containing 0.1%v/v formic acid and 20%v/v acetonitrile that can be employed for HPLC and UHPLC (Ultra High performance Liquid Chromatography) techniques. This pre-blended mobile phase solvent is of high purity that reduces time-consuming preparation and prevents instrument downtime because of impure mobile phases. Formic acid is used as mobile phase additive. Acetonitrile (MeCN), an organic solvent has low viscosity, high elution strength and is UV transparent at low wavelength.
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-05-8
Solution Contains 0.1 % (v/v) formic acid
Density 0.971 g/mL (at 25°C)
Linear Formula H2O
MDL Number MFCD00011332
Type Organic Standard
Quantity 4 x 4 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.