Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water Solution, For HPLC, Contains 0.1% (v/v) Formic Acid, MilliporeSigma™ Supelco™

Catalog No. 111005322 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
4 x 4 L
200 L
Packaging:
Pure-Pac™ 1
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
11-100-5322 200 L Pure-Pac™ 1
11-100-3684 4 x 4 L Pure-Pac™ 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-5322 Supplier MilliporeSigma Supelco Supplier No. 576913200LLS
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

The product is water containing 0.1% v/v formic acid. This pre-blended mobile phase solvent is of high purity that reduces time-consuming preparation and prevents instrument downtime because of impure mobile phases. It can be employed for HPLC and UHPLC (Ultra High performance Liquid Chromatography) techniques.

TRUSTED_SUSTAINABILITY

Specifications

Packaging Pure-Pac™ 1
Linear Formula H2O
MDL Number MFCD00011332
Quantity 200 L
Grade For HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.