Learn More
Hanna Pharmaceutical Supply Co., Inc. STERILE WATER INJ.50ML
Catalog No. NC9025748
Encompass_Preferred
50ml Fliptop Vial Sterile, nonpyrogenic, solute-free preparation of distilled water for injection. For use only as a sterile solvent or diluent vehicle for drugs or solutions suitable for parenteral administration pH 5.5 (5.0-7.0). Contains no bacteriostat, antimicrobial agent or added buffer 25/PK
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.
This item is not returnable. View return policy
Catalog No. NC9025748 Supplier Hanna Pharmaceutical Supply Co., Inc. Supplier No. 0409488750
 Restricted Item
Add to Cart
Add to Cart
50ml Fliptop Vial Sterile, nonpyrogenic, solute-free preparation of distilled water for injection. For use only as a sterile solvent or diluent vehicle for drugs or solutions suitable for parenteral administration pH 5.5 (5.0-7.0). Contains no bacteriostat, antimicrobial agent or added buffer 25/PK
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.