Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Water, Sterile, Nuclease Free, OmniPur™, Calbiochem™,

Catalog No. 98115L Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 L
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
98-115-L 5 L Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 98-115-L Supplier MilliporeSigma™ Supplier No. 98115L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recommended Storage

Ok to freeze

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Color Colorless
Grade Molecular Biology
Packaging Plastic Bottle
Quantity 5 L
Physical Form Liquid
storageNote1 Ok to freeze
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.