Learn More
Sigma Aldrich Fine Chemicals Biosciences Water sterle fltered Bo500ML
Catalog No. 502903183
Encompass
This product is a very low endotoxin (less than 1 EU/mL) 0.2 micron filtered.
Catalog No. 50-290-3183 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W350024X500ML
This product is a very low endotoxin (less than 1 EU/mL) 0.2 micron filtered.