Learn More
BINDER™ Water Supply Set
SDP
Catalog No. 08111339 Shop All BINDER Inc. Products
Change view
Click to view available options
Model:
Free-standing
Rear-mounting
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
08-111-339 Free-standing
08-111-338 Rear-mounting
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 08-111-339 Supplier BINDER™ Supplier No. 80121846
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Water Supply Set consisting of fresh- and waste-water containers (20 liters each), cabling and pump.
  • External
  • Includes fresh and waste water containers (20 L), cabling, and pump
Specifications
Description Water supply set - external, free-standing,consisting of fresh- and waste-water containers (20 liters each), cabling and pump
For Use With (Equipment) BINDER Chambers Series KBWF, KBF, KBF P, KBF LQC, MKF 56
Material Plastic
Type Water Supply
Includes Fresh- and Waste Water Containers (20 Liters Each), Cabling and Pump
Model Free-standing