Learn More
MSC Pro-Source Dehumidifier, 70 Pt. Capacity, Commercial, 1 Ea.
Catalog No. 5000509227
Encompass
This commercial-grade dehumidifier is designed to effectively manage moisture levels in environments requiring consistent humidity control. It features a 70 pt. saturation capacity and utilizes R410A refrigerant for efficient operation within a temperature range of 41°F to 89.6°F.
  • Saturation capacity of 70 pt.
  • Equipped with an 8.2 ft. power cord
  • Operates at 115 V and 60 Hz
  • Airflow rating of 221.00 cubic feet per minute
  • Power consumption of 635.00 W
  • Includes a 5 L volume capacity
  • ETL certified
Catalog No. 50-005-09227 Supplier MSC Supplier No. 98552524
This commercial-grade dehumidifier is designed to effectively manage moisture levels in environments requiring consistent humidity control. It features a 70 pt. saturation capacity and utilizes R410A refrigerant for efficient operation within a temperature range of 41°F to 89.6°F.
  • Saturation capacity of 70 pt.
  • Equipped with an 8.2 ft. power cord
  • Operates at 115 V and 60 Hz
  • Airflow rating of 221.00 cubic feet per minute
  • Power consumption of 635.00 W
  • Includes a 5 L volume capacity
  • ETL certified