Description
Lovibond™ Water Testing Minikit is easy to use and provides highly accurate results.
- Easy operation and exact reagent dosing
- High accuracy
- Foil-wrapped Lovibond™ tablet reagents with a minimum guaranteed shelf life of 5 years
- Unrestricted shipment
- Safe storage
Specifications
Specifications
| Analytes | Alkalinity Total M |
| Includes | Flip top case containing a shaker tube, rapid dissolving tablet reagents and instructions. |
| Quantity | 1 Pack |
| Range | 10 to 500 mg/L |
| Sufficient For | Approximately 40 Tests |
| Test Method | Tablet count |
| Type | Minikit water testing kit |