Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, TraceSELECT™, For Trace Analysis, Solstice

Catalog No. 60047767 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
2.5 L
10 L
4 x 2.5 L
Packaging:
Cubitainer
Glass Bottle
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-767 250 mL Poly Bottle
60-047-769 500 mL Poly Bottle
60-047-765 1 L Glass Bottle
60-047-766 2.5 L Poly Bottle
60-047-764 10 L Cubitainer
60-047-768 4 x 2.5 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 60-047-767 Supplier Solstice Supplier No. 95305250ML
Add to Cart
Add to Cart

TraceSELECT™, for trace analysis

  • High purity solvents for speciation analysis by LC-ICP-MS

  • Rigorous purification procedures followed by UV spectroscopy, IC, and ICP-MS testing to assure high chemical purity and high UV transmittance

  • The blank values for metal traces in these solvents are in the ppb range or lower.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

pH 6 to 8 (at 20°C)
Color Colorless
Melting Point 0°C
Boiling Point 100°C
UN Number NONH for all modes of transport
Beilstein 2050024
Formula Weight 18.02g/mol
Grade TraceSELECT
Odor Odorless
Packaging Poly Bottle
Density 1g/mL at 20°C
Quantity 250 mL
Physical Form Liquid
Chemical Name or Material Water
Show More Show Less
RTECSNumber 231-791-2
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.