Learn More
Med Vet International Dynarex Disposable Water Trap, Y Cap, Use w/22 mm Tubing, 60 Cc, Pediatric, 50/Case
Catalog No. 502830240
Encompass_Preferred
The Dynarex Resp-O2™ Disposable Water Trap is designed to seal off excess moisture and gasses during oxygen therapy and prevent them from backing up into tubing. The Water Trap features a threaded collection jar and self-sealing Y-shaped lid to ensure that circuits remain clear and unobstructed. Designed to trap liquids and help prevent obstructions in corrugated/circuit tubing Features threaded jar and Y-sealing lid to ensure uninterrupted flow Constructed from high-quality plastic materials for optimal reliability Not made with natural rubber latex or DEHP Available in Adult (accepts 22mm tubing) and Pediatric (accepts 15mm tubing) sizes 
Catalog No. 50-283-0240 Supplier Med Vet International Supplier No. 34203
The Dynarex Resp-O2™ Disposable Water Trap is designed to seal off excess moisture and gasses during oxygen therapy and prevent them from backing up into tubing. The Water Trap features a threaded collection jar and self-sealing Y-shaped lid to ensure that circuits remain clear and unobstructed. Designed to trap liquids and help prevent obstructions in corrugated/circuit tubing Features threaded jar and Y-sealing lid to ensure uninterrupted flow Constructed from high-quality plastic materials for optimal reliability Not made with natural rubber latex or DEHP Available in Adult (accepts 22mm tubing) and Pediatric (accepts 15mm tubing) sizes 