Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Water Treatment and Filtration
GREENER_CHOICESCI_ED

Catalog No. S97623 Shop All Aldon Products
Change view
Click to view available options
Sufficient For:
15 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
S97623 15 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Respirable Crystalline Silica, which is known to the State of California to cause cancer.
Catalog No. S97623 Supplier Innovating Science™ Supplier No. IS9700
Add to Cart
Add to Cart
WARNING: This product can expose you to chemicals including Respirable Crystalline Silica, which is known to the State of California to cause cancer.

Includes

1500 g fine sand, 1500 g coarse sand, 1500 g fine gravel, 300g activated charcoal, 25 g potassium aluminum sulfate (alum), 25 g calcium oxide (lime), 5 g Kaolin (clay) powder, 5mL green food coloring, 5mL odor agent, 75 plastic cups and 15 stirring sticks

Requires

Container large enough to prepare untreated water (≈1 gal. or larger); miscellaneous materials to prepare untreated water such as: potting soil, lawn clippings, dried and crushed leaves, small twigs, mulch, etc.; and a sharp item (such as a nail or dissecting probe) for students to poke small holes in the cups used for filtration

Develop a knowledge of the processes performed at a water treatment plant and understand the reasons for each.

  • Use water to perform on a small-scale several procedures that occur in a water treatment plant.
  • Examine the changes in the water after each treatment step is performed
  • Observe physical characteristics of water, such as clarity, color, odor and how each is affected from the beginning of the treatment process until the end

Specifications

For Use With (Application) Demonstrating several procedures that occur in a water treatment plant on polluted water
Includes 1500 g fine sand, 1500 g coarse sand, 1500 g fine gravel, 300g activated charcoal, 25 g potassium aluminum sulfate (alum), 25 g calcium oxide (lime), 5 g kaolin (clay) powder, 5mL green food coloring, 5mL odor agent, 75 plastic cups and 15 stirring sticks
Quantity 1
Sufficient For 15 Students
Type Water Treatment and Filtration Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.