Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, Ultra-Purified, HPLC Grade, Cenmed™
SDP

Catalog No. 21520119
Change view
Click to view available options
Quantity:
1 gal.
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-520-119 20 L
21-520-118 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-520-119 Supplier Cen-Med Enterprises Supplier No. 35331920L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Suitable for use as a clinical laboratory reagent. Water, Ultra-Purified, HPLC Grade; Cenmed™ is highly purified deionized water. It is packaged in solvent-rinsed amber glass bottles and sealed under an inert atmosphere for the highest degree of quality control.

This ultra-purified, deionized water is the ideal product for use in laboratory applications and is suitable for gradient and isocratic conditions. It is free from organic and inorganic compounds and has no UV absorbance.

  • HPLC-grade laboratory reagent
  • Available in 20L and 1 gallon quantities
  • Packaged in plastic containers

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

pH 7
Color Colorless
Melting Point 0°C
Boiling Point 100°C
Formula Weight 18.02g/mol
Grade HPLC
Packaging Plastic
Quantity 20 L
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.