Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, USP/EP Purified, Ricca Chemical
SDP

Catalog No. R91900004F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
4 L
10 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
Hard Pack
Natural Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R91900004F 4 L Cubitainer
9190-1G 4 L Amber Glass Bottle
9190-1 4 L Natural Poly Bottle
9190-25 10 L Cubitainer
9190-5 20 L Cubitainer
91-905-HP 20 L Hard Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. R91900004F Supplier Ricca Chemical Company Supplier No. R91900004F
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Water, USP/EP Purified

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
PubChem CID 962
ChEBI CHEBI:15377
SMILES O

Specifications

pH 5.0 to 7.0
Color Clear, Colorless
CAS Min % 98.0
CAS Max % 100.0
Conductivity max. 1 μS/cm
Endotoxin max. 0.25 EU/mL at time of manufacture
Synonym dihydrogen oxide
IUPAC Name water
Molecular Weight (g/mol) 18.02
Percent Purity 100%
Grade EP/USP
Packaging Cubitainer
Residue after Evaporation max. 0.001 %
Sterility Non-sterile
Quantity 4 L
Chemical Name or Material Water
Show More Show Less
ShelfLife 12 Months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.