Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, USP Purified, Nonsterile, Ricca Chemical
SDP

Catalog No. R9191000330 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
55 gal.
330 gal.
Packaging:
Poly Drum
Tote
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R9191000330 330 gal. Tote
919100055E 55 gal. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R9191000330 Supplier Ricca Chemical Company Supplier No. R9191000330E3
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES O

Specifications

pH 5 to 7
Color Colorless
CAS Max % 100
Conductivity max. 1 μS/cm
IUPAC Name water
Molecular Weight (g/mol) 18.02
Percent Purity 100%
Grade USP Purified
Packaging Tote
Quantity 330 gal.
Chemical Name or Material Water
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.