Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Water, WFI Quality, Sterile Purified Water, Cell Culture Tested, OmniPur™, Calbiochem™,

Catalog No. 4865059200 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
1 L
5 L
10 L
20 L
200 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-650-59200 200 L
48-650-50500 500 mL
48-650-51000 1 L
48-650-55000 5 L
48-650-59010 10 L
48-650-59020 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 48-650-59200 Supplier MilliporeSigma™ Supplier No. 4.86505.9200
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Color Colorless
Packaging Plastic Drum
Quantity 200 L
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.