Learn More
Research Products International Corp WaterBath Clear Algae Inhibitor, (480ml) 16 Ounces
SDP
Catalog No. 50550227
Encompass_Preferred
WaterBath Clear contains an active organic ingredient that inhibits Algae growth in water baths and incubators. Ensures crystal clear water and prevents algae growth. Keeps the water bath free from algae, reducing the need for frequent cleaning and maintenance. By preventing algae buildup, it ensures that the water bath operates at peak efficiency, providing consistent and reliable temperature control for experiments. Algae Inhibitor reduces the risk of contamination, ensuring that your samples are maintained in a clean environment, which is crucial for accurate and reproducible results.
Simply add 0.5ml per liter of water to effectively treat water. Economical to use. Eight ounces treats approximately 500 liters of water.
Catalog No. 50-550-227 Supplier Research Products International Corp Supplier No. 200122
WaterBath Clear contains an active organic ingredient that inhibits Algae growth in water baths and incubators. Ensures crystal clear water and prevents algae growth. Keeps the water bath free from algae, reducing the need for frequent cleaning and maintenance. By preventing algae buildup, it ensures that the water bath operates at peak efficiency, providing consistent and reliable temperature control for experiments. Algae Inhibitor reduces the risk of contamination, ensuring that your samples are maintained in a clean environment, which is crucial for accurate and reproducible results.
Simply add 0.5ml per liter of water to effectively treat water. Economical to use. Eight ounces treats approximately 500 liters of water.