Learn More
GRANT INSTRUMENTS Water bath, digital, 18L ambient +5 to 99°C, includes clear lid, drain and base tray
SDP
Catalog No. NC2236242
Encompass_Preferred
Built to the highest standard and specifications and incorporating the latest technology the SUB Aqua Pro advanced water bath range supports even the most demanding applications requiring accurate temperature control.A choice of eight models with a base tray and lid included as standard.Temperature range ambient +5ºC to 99ºC operation.Stability: ±0.2ºC.Unique Set and Forget technology - fast heat-up, accurate temperature control.Simple, intuitive controls - quick and easy to set temperature.Suitable for use with heat transfer beads (excluding SAP2 & SAP2S).Adjustable over temperature alarm - protect samples from over heating.Practical front panel lock - disables front panel controls preventing unintentional changes.Applications:Sample preparation, sample incubation, sample warming, sample thawing, media preparation, QC materials and practical science demonstration.Markets: Pharma/biotech, education, industry, healthcare.
This item is not returnable. View return policy
Catalog No. NC2236242 Supplier GRANT INSTRUMENTS Supplier No. SAP18 US
Built to the highest standard and specifications and incorporating the latest technology the SUB Aqua Pro advanced water bath range supports even the most demanding applications requiring accurate temperature control.A choice of eight models with a base tray and lid included as standard.Temperature range ambient +5ºC to 99ºC operation.Stability: ±0.2ºC.Unique Set and Forget technology - fast heat-up, accurate temperature control.Simple, intuitive controls - quick and easy to set temperature.Suitable for use with heat transfer beads (excluding SAP2 & SAP2S).Adjustable over temperature alarm - protect samples from over heating.Practical front panel lock - disables front panel controls preventing unintentional changes.Applications:Sample preparation, sample incubation, sample warming, sample thawing, media preparation, QC materials and practical science demonstration.Markets: Pharma/biotech, education, industry, healthcare.