Learn More
Cole-Parmer Winland Electronics WB200 Wb 200 Waterbug Alert Console HardWire 12 or 24 V or VDC
Catalog No. NC1909618 Shop All Cole-Parmer Products
Encompass
WaterBug products provide reliable water detection while not alarming due to high humidity or condensation. When integrated with an alarm system, these units will activate dialers and alarms to notify of a problem. Optional add-ons include a 12V power supply for stand-alone operation and a remote annunciator for remote alarm notification in an additional location. All units utilize standard surface and standard under-carpet sensors; however the WB800 can also support supervised sensors that alarm if water is detected as well as if a lead is severed. Designed and manufactured in USA. WB350 - Stand-Alone WaterBug Alert Console Designed for stand-alone operation powered by a 9V battery with low battery indicator Ideal for office environments or near water heaters
Catalog No. NC1909618 Supplier Cole-Parmer Supplier No. 8955177
WaterBug products provide reliable water detection while not alarming due to high humidity or condensation. When integrated with an alarm system, these units will activate dialers and alarms to notify of a problem. Optional add-ons include a 12V power supply for stand-alone operation and a remote annunciator for remote alarm notification in an additional location. All units utilize standard surface and standard under-carpet sensors; however the WB800 can also support supervised sensors that alarm if water is detected as well as if a lead is severed. Designed and manufactured in USA. WB350 - Stand-Alone WaterBug Alert Console Designed for stand-alone operation powered by a 9V battery with low battery indicator Ideal for office environments or near water heaters