Learn More
Description
Thermo Scientific™ WaterBugs Klebsiella pneumoniae Strain C6 is used for water/wastewater quality control testing; microorganisms are supplied in a Quanti-Cult format.
- Deliver <50 CFUs in a single inoculum to mimic water samples
- One test/reconstituted vial
Ordering Alerts:
Supplied with 10 preserved organism vials; 10 rehydration vials.
Specifications
Specifications
| Description | Klebsiella pneumoniae Strain C6 |
| Format | Quanti-Cult |
| Product Type | Quality Control Organism |
| Target Organism Class | Gram-negative Rods |
| Test Type | Quantitative QC |
| Packaging Type | 10 Test/Kit |
| Quantity | 10 Tests/Kit |
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.