Learn More
GFS Chemicals Inc WATERMARK KARL FISCHER TITRANT, STABLE, 5 mg/ml, NON-HYGROSCOPIC | 1 L | GFS CHEMICALS, INC. Item #: 99661 | Bulk #: 1604
SDP
Catalog No. NC1625571
Encompass_Preferred
WATERMARK KARL FISCHER TITRANT, STABLE, 5 mg/ml, NON-HYGROSCOPIC | For volumetric analysis. Must be used in conjunction with KF Solvent 1609, 1610 or Buffer 1615. ; Specifications : Titer strength (mg/ml) 4.95 - 5.05 mg/ml
Catalog No. NC1625571 Supplier GFS Chemicals Inc Supplier No. 99661
WATERMARK KARL FISCHER TITRANT, STABLE, 5 mg/ml, NON-HYGROSCOPIC | For volumetric analysis. Must be used in conjunction with KF Solvent 1609, 1610 or Buffer 1615. ; Specifications : Titer strength (mg/ml) 4.95 - 5.05 mg/ml