Learn More
GFS Chemicals Inc WATERMARK KARL FISCHER WATER STANDARD, 1.0 mg/g | 10 X 5 ML | GFS CHEMICALS, INC. Item #: 99862 | Bulk #: 2302
SDP
Catalog No. NC1439691
Encompass_Preferred
WATERMARK KARL FISCHER WATER STANDARD, 1.0 mg/g | Water standard for Karl Fischer standardization. ; Specifications : NIST Traceable, Water (mg/g) 0.97 - 1.03 mg/g
Catalog No. NC1439691 Supplier GFS Chemicals Inc Supplier No. 99862
WATERMARK KARL FISCHER WATER STANDARD, 1.0 mg/g | Water standard for Karl Fischer standardization. ; Specifications : NIST Traceable, Water (mg/g) 0.97 - 1.03 mg/g