Learn More
GFS Chemicals Inc WATERMARK KARL FISCHER COULOMETRIC GENERATOR SOLUTION, UNIVERSAL | 10 X 5 ML | GFS CHEMICALS, INC. Item #: 99713 | Bulk #: 2321
SDP
Catalog No. NC1922204
Encompass_Preferred
WATERMARK KARL FISCHER COULOMETRIC GENERATOR SOLUTION, UNIVERSAL | Catholyte solution for coulometric analysis. For both general use and aldehydes and ketones. ; Specifications : Moisture (ppm) 50 ppm , Suitability
Catalog No. NC1922204 Supplier GFS Chemicals Inc Supplier No. 99713
WATERMARK KARL FISCHER COULOMETRIC GENERATOR SOLUTION, UNIVERSAL | Catholyte solution for coulometric analysis. For both general use and aldehydes and ketones. ; Specifications : Moisture (ppm) 50 ppm , Suitability