Learn More
GFS Chemicals Inc WATERMARK KARL FISCHER SOLVENT, GENERAL PURPOSE | 1 L | GFS CHEMICALS, INC. Item #: 99741 | Bulk #: 1610
SDP
Catalog No. NC0796965
Encompass_Preferred
WATERMARK KARL FISCHER SOLVENT, GENERAL PURPOSE | For volumetric analysis. To be used with any GFS KF Titrant for general purpose use. Not for testing aldehydes and ketones.
Catalog No. NC0796965 Supplier GFS Chemicals Inc Supplier No. 99741
WATERMARK KARL FISCHER SOLVENT, GENERAL PURPOSE | For volumetric analysis. To be used with any GFS KF Titrant for general purpose use. Not for testing aldehydes and ketones.