Learn More
Thermco Products Inc WATERPF DIG THERMOM 50/300C
Catalog No. NC1121622
Encompass_Preferred
NIST Traceable w/Certificate5”x 0.14” Stainless Steel Stem w/Probe Cover.Head 1 3/4”dia.Water Resistant.Shock proof.Minimum/Maximum Memory.Hold/Test Function.C/F SwitchableRange: -50°/300°C (-58°/572°F)Accuracy: ± 1.0ºC between -20°to 120°CResolution: -20° to 200° = 0.1°, outside range 1.0°
Catalog No. NC1121622 Supplier Thermco Products Inc Supplier No. ACC400DIG
Add to Cart
Add to Cart
NIST Traceable w/Certificate5”x 0.14” Stainless Steel Stem w/Probe Cover.Head 1 3/4”dia.Water Resistant.Shock proof.Minimum/Maximum Memory.Hold/Test Function.C/F SwitchableRange: -50°/300°C (-58°/572°F)Accuracy: ± 1.0ºC between -20°to 120°CResolution: -20° to 200° = 0.1°, outside range 1.0°