Learn More
Deltatrak Inc WATERPRF LOLLIPOP THERMOMETER, Packaged individually
SDP
Catalog No. NC0238859
Encompass_Preferred
The Waterproof Lollipop, Min/Max Digital Thermometer (Model 11050) is ideal for commercial dishwashers. With the thermometers auto-calibration and min/max features, users can quickly verify and quantify the final rinse temperature in commercial warewashers and dishwashers. Measurement Range -40F to 311F (-40C to 155C) Operating Range 32F to 122F (0C to 50C) Accuracy 1F (14F to 194F), 0.5C (-10C to 90C) or 1% whichever is greatest Sensor Thermistor Display Resolution 0.1F /C Display Reading Update 1 second Display Window Size 1.3 in W x 0.9 in H (32.9 mm W x 22.9 mm H) Calibration Auto calibration feature Battery Button Cell Alkaline Body Casing ABS plastic Probe Length 4.13 in. (105 mm) Probe Diameter 0.14 in. (3.56 mm), reduced tip 0.1 in (2.5mm) Weight 1.3 oz (37.6g) Waterproof Rating IP65 Factory Calibration NIST traceable calibration certified Compliance Certificate Manufacturers certificate of compliance available from DeltaTrak, NIST Traceable, NSF, CE
Catalog No. NC0238859 Supplier Deltatrak Inc Supplier No. 11050
The Waterproof Lollipop, Min/Max Digital Thermometer (Model 11050) is ideal for commercial dishwashers. With the thermometers auto-calibration and min/max features, users can quickly verify and quantify the final rinse temperature in commercial warewashers and dishwashers. Measurement Range -40F to 311F (-40C to 155C) Operating Range 32F to 122F (0C to 50C) Accuracy 1F (14F to 194F), 0.5C (-10C to 90C) or 1% whichever is greatest Sensor Thermistor Display Resolution 0.1F /C Display Reading Update 1 second Display Window Size 1.3 in W x 0.9 in H (32.9 mm W x 22.9 mm H) Calibration Auto calibration feature Battery Button Cell Alkaline Body Casing ABS plastic Probe Length 4.13 in. (105 mm) Probe Diameter 0.14 in. (3.56 mm), reduced tip 0.1 in (2.5mm) Weight 1.3 oz (37.6g) Waterproof Rating IP65 Factory Calibration NIST traceable calibration certified Compliance Certificate Manufacturers certificate of compliance available from DeltaTrak, NIST Traceable, NSF, CE