Learn More
Enterprise Technology Solutions Bluetooth Waterproof Barcode Scanner
SDP
Catalog No. 502111224
Encompass_Preferred
The NuScan 4100B Bluetooth Handheld CCD Barcode Scanner from Adesso delivers remarkable performance and versatility for industrial environments. It offers a wide scanning range for multiple symbologies from up to 30’ away. The scanner withstands accidental drops from up to 20' and is standard IP67-rated water and dust resistant. It can easily scan your barcode from up to a foot away.
Catalog No. 50-211-1224 Supplier Enterprise Technology Solutions Supplier No. NUSCAN4100B
The NuScan 4100B Bluetooth Handheld CCD Barcode Scanner from Adesso delivers remarkable performance and versatility for industrial environments. It offers a wide scanning range for multiple symbologies from up to 30’ away. The scanner withstands accidental drops from up to 20' and is standard IP67-rated water and dust resistant. It can easily scan your barcode from up to a foot away.