Learn More
Grainger Waterproof Cable Assembly CPVC BNC
Catalog No. 502767964
Encompass_Preferred
The SensoRex Submersible Electrode is a waterproof pH electrode designed for continuous monitoring and control applications. It has a gel-filled epoxy body, double junction reference, built-in temperature sensor, and 3/4" NPT mounting thread. The electrode measures pH from 0-14 pH with ±0.1 pH accuracy. It has a response time of <30 seconds and operating temperature range of 0-80°C. The electrode requires a BNC connector and is compatible with the SensoRex SX series transmitters. It has a fully submersible IP68 rated body for use in tanks, pipes, or open vessels in chemical, water treatment, agriculture, aquaculture, and other applications requiring continuous pH measurement.
Catalog No. 50-276-7964 Supplier Grainger Supplier No. 16Z018
The SensoRex Submersible Electrode is a waterproof pH electrode designed for continuous monitoring and control applications. It has a gel-filled epoxy body, double junction reference, built-in temperature sensor, and 3/4" NPT mounting thread. The electrode measures pH from 0-14 pH with ±0.1 pH accuracy. It has a response time of <30 seconds and operating temperature range of 0-80°C. The electrode requires a BNC connector and is compatible with the SensoRex SX series transmitters. It has a fully submersible IP68 rated body for use in tanks, pipes, or open vessels in chemical, water treatment, agriculture, aquaculture, and other applications requiring continuous pH measurement.