Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Waterproof Elbow Length Cryogenic Gloves
SDP

Catalog No. 17355065 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-355-065 Medium
17-355-066 Large
17-355-067 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 17-355-065 Supplier National Safety Apparel™ Supplier No. G99CRBEPMDEL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Gloves feature a breathable membrane for additional splash protection.

  • Teflon™ Laminated Nylon with 3M™ Scotchlite™ Thinsulate™ liner
  • Waterproof and breathable membrane for additional splash protection
  • Made in the USA

Specifications

Certifications/Compliance (Europe) EN 511 Contact Cold Performance Level 2, EN 511 Convective Cold Performance Level 3
Chlorinated No
Description Elbow Length
Color Blue
Exterior Finish Smooth
Hand Specific
Material Nylon
Product Type Cryogenic Gloves
Size Medium
For Use With (Application) Cryogenic Liquids/Gases
Glove Cuff Style Slip-on
Liner Material 3M™ Thinsulate™
Lining Lined
Series G99CRBEP_ _EL
Length (English) 18 in.
Length (Metric) 45.7 cm
Product Line Waterproof Cryogenic Gloves
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.