Learn More
Cole-Parmer Cole-Parmer Solution Label w/Waterproof Overlay, 2"L x 1" H; 1000/RL
Catalog No. NC9426412 Shop All Cole-Parmer Products
Encompass_Preferred
Cole-Parmer Solution Label w/Waterproof Overlay, 2"L x 1" H; 1000/RL, Use these labels to organize your samples. Fill out the label using a pen, pencil, or marker; plastic overlay permanently protects the label. Pressure-sensitive. Roll of 1000.
Catalog No. NC9426412 Supplier Cole-Parmer Supplier No. 6220400
Cole-Parmer Solution Label w/Waterproof Overlay, 2"L x 1" H; 1000/RL, Use these labels to organize your samples. Fill out the label using a pen, pencil, or marker; plastic overlay permanently protects the label. Pressure-sensitive. Roll of 1000.