Learn More
World Precision Instrument Waterpump Conversion Kit
SDP
Catalog No. 502538838
Encompass
Set of male and female quick disconnects and clamps to convert standard water heating pump for use with EZ Anesthesia water-heated surgical beds.
Catalog No. 50-253-8838 Supplier World Precision Instrument Supplier No. EZ2002
Set of male and female quick disconnects and clamps to convert standard water heating pump for use with EZ Anesthesia water-heated surgical beds.