Learn More
Waters Corp Waters Vial Holder, Rectangular, 24-position
Supplier:  Waters Corp 405000560
Encompass_Preferred
This rectangular vial-holder plate is designed to accommodate either 2 mL autosampler vials or 1.5 mL microcentrifuge tubes.
  • Has 24 positions for sample handling
  • Accommodates 2 mL autosampler vials
  • Accommodates 1.5 mL microcentrifuge tubes (Eppendorf style)
  • Designed for use with 2790/2795 Separations Module
Catalog No. 50-791-745