Learn More
Waters Corp Waters Sample Loop, UPLC, Stainless Steel, 20 μL
Catalog No. NC9010978 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This sample loop is designed for high-performance liquid chromatography systems. It fits into the 5000psi nanoACQUITY system and is suitable for precise sample injection.
  • Constructed from stainless steel
  • Volume of 20 μL
  • Compatible with UPLC systems
  • Designed for nanoACQUITY systems
  • Replacement as required
Catalog No. NC9010978 Supplier Waters Corp Supplier No. 430001320
This sample loop is designed for high-performance liquid chromatography systems. It fits into the 5000psi nanoACQUITY system and is suitable for precise sample injection.
  • Constructed from stainless steel
  • Volume of 20 μL
  • Compatible with UPLC systems
  • Designed for nanoACQUITY systems
  • Replacement as required