Learn More
Analytical Sales & Services Waters Alliance 2690/2695 & 27
Catalog No. 5000806099
Encompass
Waters Alliance 2690/2695 & 2790/2795/2796 Direct Replacement PEEK Inlet or Outlet Cartridge OEM No. 270941\n\nQty/Pkg: EACH
Catalog No. 50-008-06099 Supplier Analytical Sales & Services Supplier No. 925170
Waters Alliance 2690/2695 & 2790/2795/2796 Direct Replacement PEEK Inlet or Outlet Cartridge OEM No. 270941\n\nQty/Pkg: EACH