Learn More
Waters Corp Waters HPLC Absorbance PQ Test Solution, UV Detection, Caffeine Standards, 10 mL
Catalog No. 50466683 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This solution, for HPLC systems with UV detection, calibrates and troubleshoots UV/Vis absorbance detectors. It ensures repeatable absorbance values, identifies issues like misaligned optics or dirty flow cells, and is ready-to-use for LC analyses and troubleshooting.
  • Meticulously formulated reference solution calibrates and troubleshoots UV/Vis absorbance detectors.
  • Contains precise light-absorbing compounds for verifying instrument settings, identifying detector issues, and ensuring known, repeatable absorbance values.
  • Pinpoints detector problems such as misaligned optics or dirty flow cells.
  • Ready-to-use format for convenience, suitable for diverse LC applications including quantitative and qualitative analyses.
  • Useful for troubleshooting LC instrument challenges.
  • Prioritizes safety and ease of use.
  • Available in various packaging options.
Catalog No. 50-466-683 Supplier Waters Corp Supplier No. 700005906
This solution, for HPLC systems with UV detection, calibrates and troubleshoots UV/Vis absorbance detectors. It ensures repeatable absorbance values, identifies issues like misaligned optics or dirty flow cells, and is ready-to-use for LC analyses and troubleshooting.
  • Meticulously formulated reference solution calibrates and troubleshoots UV/Vis absorbance detectors.
  • Contains precise light-absorbing compounds for verifying instrument settings, identifying detector issues, and ensuring known, repeatable absorbance values.
  • Pinpoints detector problems such as misaligned optics or dirty flow cells.
  • Ready-to-use format for convenience, suitable for diverse LC applications including quantitative and qualitative analyses.
  • Useful for troubleshooting LC instrument challenges.
  • Prioritizes safety and ease of use.
  • Available in various packaging options.