Learn More
Analytical Sales & Services Waters Models 2690/2695 & 2790 Wash Tube Seal
Catalog No. 5000806541
Encompass
Waters Models 2690/2695 & 2790/2795/2796 Replacement UHMWPE Wash Tube Seal OEM No. 270940\n\nQty/Pkg: EACH
Catalog No. 50-008-06541 Supplier Analytical Sales & Services Supplier No. 925214
Waters Models 2690/2695 & 2790/2795/2796 Replacement UHMWPE Wash Tube Seal OEM No. 270940\n\nQty/Pkg: EACH