Learn More
Analytical Sales & Services Waters Models 45 45G M501 5
Catalog No. 5000804868
Encompass
Waters Models 45 45G M501 510 515 590 600 600E 610 1515 1525 (Analtyical) 6000 6000A & LCM1 Self-Priming Outlet Check Valve Cartridge Only\n\nQty/Pkg: EACH
Catalog No. 50-008-04868 Supplier Analytical Sales & Services Supplier No. 925005
Waters Models 45 45G M501 510 515 590 600 600E 610 1515 1525 (Analtyical) 6000 6000A & LCM1 Self-Priming Outlet Check Valve Cartridge Only\n\nQty/Pkg: EACH