Learn More
Analytical Sales & Services Waters Models 501 600 600E &
Catalog No. 5000805051
Encompass
Waters Models 501 600 600E & 610. Self-Priming Outlet Check Valve Assembly Includes Cartridge and Housing OEM No.025216/700000253\n\nQty/Pkg: EACH
Catalog No. 50-008-05051 Supplier Analytical Sales & Services Supplier No. 925027
Waters Models 501 600 600E & 610. Self-Priming Outlet Check Valve Assembly Includes Cartridge and Housing OEM No.025216/700000253\n\nQty/Pkg: EACH