Learn More
Analytical Sales & Services Waters Models 616 Stainless St
Catalog No. 5000804235
Encompass
Waters Models 616 Stainless Steel Self-Priming Outlet Check Valve Assembly Includes Cartridge and Housing OEM No. 55845 & 24960\n\nQty/Pkg: EACH
Catalog No. 50-008-04235 Supplier Analytical Sales & Services Supplier No. 925062
Waters Models 616 Stainless Steel Self-Priming Outlet Check Valve Assembly Includes Cartridge and Housing OEM No. 55845 & 24960\n\nQty/Pkg: EACH