Learn More
Analytical Sales & Services Waters Models 625 & 626 PEEK
Catalog No. 5000806782
Encompass
Waters Models 625 & 626 PEEK Direct Replacement Self-Priming Inlet Check Valve Cartridge Only OEM No. 02412\n\nQty/Pkg: EACH
Catalog No. 50-008-06782 Supplier Analytical Sales & Services Supplier No. 925110
Waters Models 625 & 626 PEEK Direct Replacement Self-Priming Inlet Check Valve Cartridge Only OEM No. 02412\n\nQty/Pkg: EACH