Learn More
Analytical Sales & Services Waters Models M45 45G 501 5
Catalog No. 5000805804
Encompass
Waters Models M45 45G 501 510 515 590 600 600E 610 1515 1525 (Analtyical) 6000 6000A & LCM1 Replacement UHMWPE Piston Seal with Teflon Gasket OEM No. 022946/022934 10/PK\n\nQty/Pkg: 10PK
Catalog No. 50-008-05804 Supplier Analytical Sales & Services Supplier No. 925103
Waters Models M45 45G 501 510 515 590 600 600E 610 1515 1525 (Analtyical) 6000 6000A & LCM1 Replacement UHMWPE Piston Seal with Teflon Gasket OEM No. 022946/022934 10/PK\n\nQty/Pkg: 10PK