Learn More
Analytical Sales & Services Waters Models M45 45G 501 5
Catalog No. 5000805610
Encompass
Waters Models M45 45G 501 510 515 590 600 600E 610 1515 1525 (Analtyical) 6000 6000A & LCM1 Replacement Teflon Piston Seal with Teflon Gasket OEM No. 026613 10/PK\n\nQty/Pkg: 10PK
Catalog No. 50-008-05610 Supplier Analytical Sales & Services Supplier No. 900313T
Waters Models M45 45G 501 510 515 590 600 600E 610 1515 1525 (Analtyical) 6000 6000A & LCM1 Replacement Teflon Piston Seal with Teflon Gasket OEM No. 026613 10/PK\n\nQty/Pkg: 10PK