Learn More
Analytical Sales & Services Waters Models M45 45G 510 5
Catalog No. 5000806266
Encompass
Waters Models M45 45G 510 515 590 1515 1525 (Analytical) 6000 6000A & LCM1 Self-Priming Outlet Check Valve Assembly Includes Cartridge and Housing OEM No. 025216/700000253\n\nQty/Pkg: EACH
Catalog No. 50-008-06266 Supplier Analytical Sales & Services Supplier No. 925025
Waters Models M45 45G 510 515 590 1515 1525 (Analytical) 6000 6000A & LCM1 Self-Priming Outlet Check Valve Assembly Includes Cartridge and Housing OEM No. 025216/700000253\n\nQty/Pkg: EACH