Learn More
Analytical Sales & Services Waters Models M45 45G M501
Catalog No. 5000804253
Encompass
Waters Models M45 45G M501 510 515 590 600. 600E 610 1515 1525 (Analytical) 6000 600A & LCM1 Self-Priming Inlet Check Valve Assembly Includes Cartridge and Housing OEM Nos. 033679/700000253\n\nQty/Pkg: EACH
Catalog No. 50-008-04253 Supplier Analytical Sales & Services Supplier No. 925020
Waters Models M45 45G M501 510 515 590 600. 600E 610 1515 1525 (Analytical) 6000 600A & LCM1 Self-Priming Inlet Check Valve Assembly Includes Cartridge and Housing OEM Nos. 033679/700000253\n\nQty/Pkg: EACH