Learn More
Analytical Sales & Services Waters Models M501 600 600E
Catalog No. 5000806268
Encompass
Waters Models M501 600 600E & 610 & Extended Flow 225µL Head #3 Models M600EF 600E EF 610EF 650EF Replacement Adapter Seals for Outlet Check Valve 5/PK\n\nQty/Pkg: 5PK
Catalog No. 50-008-06268 Supplier Analytical Sales & Services Supplier No. 925037
Waters Models M501 600 600E & 610 & Extended Flow 225µL Head #3 Models M600EF 600E EF 610EF 650EF Replacement Adapter Seals for Outlet Check Valve 5/PK\n\nQty/Pkg: 5PK