Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters MS Leucine Enkephalin Kit, 5 mL, Reference, System Performance

Catalog No. 50792756 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This multi-component kit is designed for Mass Spectrometers, providing standards and calibrants for system performance tests. It supports MS time-of-flight installation, ESI positive and negative, and APCI sensitivity and precision tests. The kit helps establish, monitor, and reproduce instrument performance, covering setup, calibration, assay assessment, and daily competence.

  • 5 mL 2ng/μL Leucine Enkephalin Solution in 50/50 Acetonitrile-water ampule.
  • 5 mL 1% Acetic Acid Solution in 50/50 Acetonitrile/water ampule.
  • 12 mL empty screw thread amber vial with cap.
  • Certificate of Analysis included.
  • Separation mode: Reversed Phase.
  • Detection method: MS.
  • Formulation: Liquid.
  • Storage: Ambient (shipping), -20 ±10°C (Freezer).
  • Shelf life: 12 Months.
  • Units per package: 1 pk.
  • Hazardous material requiring special freight handling.

Catalog No. 50-792-756 Supplier Waters Corp Supplier No. 700002456
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This multi-component kit is designed for Mass Spectrometers, providing standards and calibrants for system performance tests. It supports MS time-of-flight installation, ESI positive and negative, and APCI sensitivity and precision tests. The kit helps establish, monitor, and reproduce instrument performance, covering setup, calibration, assay assessment, and daily competence.

  • 5 mL 2ng/μL Leucine Enkephalin Solution in 50/50 Acetonitrile-water ampule.
  • 5 mL 1% Acetic Acid Solution in 50/50 Acetonitrile/water ampule.
  • 12 mL empty screw thread amber vial with cap.
  • Certificate of Analysis included.
  • Separation mode: Reversed Phase.
  • Detection method: MS.
  • Formulation: Liquid.
  • Storage: Ambient (shipping), -20 ±10°C (Freezer).
  • Shelf life: 12 Months.
  • Units per package: 1 pk.
  • Hazardous material requiring special freight handling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.