Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Prep 15/30 SFC System Test Mix and Internal Standard

Catalog No. 50466668 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Standards Kit for Waters Prep 15/30 SFC system assessment.
1 60mL container and 1 125mL container with dry compounds consisting of:
Solution 1 (60mL) Qty 1 0.3500 mg/mL (21mg) Antipyrin 0.5%, 0.7500 mg/mL (45mg) Ketoprofen 0.5% , 0.2500 mg/mL (15mg) Sulfamethazine 0.5%
Solution 2 (125mL) Qty 1 1.0000 mg/mL (125mg) Acetazolamide 0.5%
Purity of all materials should be best grade available (min >98%) unless otherwise specified

Catalog No. 50-466-668 Supplier Waters Corp Supplier No. 700005675
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Standards Kit for Waters Prep 15/30 SFC system assessment.
1 60mL container and 1 125mL container with dry compounds consisting of:
Solution 1 (60mL) Qty 1 0.3500 mg/mL (21mg) Antipyrin 0.5%, 0.7500 mg/mL (45mg) Ketoprofen 0.5% , 0.2500 mg/mL (15mg) Sulfamethazine 0.5%
Solution 2 (125mL) Qty 1 1.0000 mg/mL (125mg) Acetazolamide 0.5%
Purity of all materials should be best grade available (min >98%) unless otherwise specified

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.