Learn More
Analytical Sales & Services Waters Replacement Sapphire Ro
Catalog No. 5000805608
Encompass
Waters Replacement Sapphire Rod Piston for Models M510 515 590 600 600E 610 1515 1525 (Analytical) 6000 6000A & LCM1 OEM No. 025656\n\nQty/Pkg: EACH
Catalog No. 50-008-05608 Supplier Analytical Sales & Services Supplier No. 900201
Waters Replacement Sapphire Rod Piston for Models M510 515 590 600 600E 610 1515 1525 (Analytical) 6000 6000A & LCM1 OEM No. 025656\n\nQty/Pkg: EACH