Learn More
Analytical Sales & Services Waters Replacement Sapphire Rod Piston
Catalog No. 5000806349
Encompass
Waters Replacement Sapphire Rod Piston for Models Alliance 2690/2695 & 2790/2795/2796 OEM No. 270959\n\nQty/Pkg: EACH
Catalog No. 50-008-06349 Supplier Analytical Sales & Services Supplier No. 925145
Waters Replacement Sapphire Rod Piston for Models Alliance 2690/2695 & 2790/2795/2796 OEM No. 270959\n\nQty/Pkg: EACH