Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Spherisorb C1 Column, 80Å, 3 μm, 4.6 mm x 150 mm

Supplier:  Waters Corp PSS832913

Encompass_Preferred

Spherisorb C1 columns are classified as USP L13 and are general purpose columns that contain trimethylsilane bonded to silica.

  • C1 chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range of 2 to 8
  • 6000 psi (415 Bar) maximum pressure
  • Not endcapped
  • High silanol activity
  • Spherical particle shape
  • 3 μm particle size
  • Parker-style endfitting type
  • 80 Å pore size
  • 220 surface area
  • Designed for HPLC systems
  • 4.6 mm inner diameter and 150 mm length
  • 2% carbon load
  • 1 unit per package

Catalog No. 50-784-459


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.